Learn More
MSC REED Instruments Photo Tachometer, Non-Contact, 2 to 99,999 RPM, ±0.05% Accuracy
Catalog No. 5000513168
Encompass
This compact photo tachometer measures the RPM of rotating objects and revolution count of moving objects without contact. It features a built-in laser pointer for identifying the target area, a durable double molded plastic housing, and a backlit LCD display. The internal memory recalls maximum, minimum, and last stored values, and it includes a low battery indicator and auto shut-off function.
  • Measures RPM of rotating objects without contact
  • Counts moving objects
  • Built-in laser pointer
  • Durable double molded plastic housing
  • Backlit LCD display
  • Internal memory recalls maximum, minimum, and last stored values
  • Low battery indicator
  • Auto shut-off function
  • Includes reflective tape
  • Includes soft carrying case
  • Includes batteries
Catalog No. 50-005-13168 Supplier MSC Supplier No. 10165751
This compact photo tachometer measures the RPM of rotating objects and revolution count of moving objects without contact. It features a built-in laser pointer for identifying the target area, a durable double molded plastic housing, and a backlit LCD display. The internal memory recalls maximum, minimum, and last stored values, and it includes a low battery indicator and auto shut-off function.
  • Measures RPM of rotating objects without contact
  • Counts moving objects
  • Built-in laser pointer
  • Durable double molded plastic housing
  • Backlit LCD display
  • Internal memory recalls maximum, minimum, and last stored values
  • Low battery indicator
  • Auto shut-off function
  • Includes reflective tape
  • Includes soft carrying case
  • Includes batteries