Learn More
Aviva Systems Biology Corp REEP1 Antibody - middle region (ARP49770_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49770_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REEP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human REEP1. Alternate names: C2orf23, FLJ13110, SPG31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60175