Learn More
World Precision Instrument Dri-Ref Reference Electrode, 2 mm
SDP
Catalog No. 50821846
Encompass
Low electrolyte leakage, stable potential with low resistance
Dri-Ref™ reference electrodes were developed by WPI to have extremely low electrolyte leak­age properties, hence the name “Dri-Ref.” In addition to this key feature, these electrodes exhibit stable and reproducible potential and low resistance. Stored in KCl when not in use, they have a long life expectancy.
Catalog No. 50-821-846 Supplier World Precision Instrument Supplier No. DRIREF2
Low electrolyte leakage, stable potential with low resistance
Dri-Ref™ reference electrodes were developed by WPI to have extremely low electrolyte leak­age properties, hence the name “Dri-Ref.” In addition to this key feature, these electrodes exhibit stable and reproducible potential and low resistance. Stored in KCl when not in use, they have a long life expectancy.