Learn More
Hach Company Radiometer Analytical XR820 Reference Electrode (Ag/AgCl reference, ground joint, screw cap)
Supplier:  Hach Company B20C820
Encompass
The Radiometer XR820 Reference Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (sat. KCl + sat. AgCl upper reservoir), ground joint, and fiber and porous pin junctions. The XR820 reference probe is recommended for impedance applications. The XR820 is a screw cap electrode and requires an external connection cable for connection to a meter.
Catalog No. 50-199-7536
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.