Learn More
Fireflysci UV/VIS Photometric Accuracy & Stray Light Calibration Kit 1 (HF-0.2, HF-0.5, HF-1, WAV-1 UV/VIS, Blank Holder)
SDP
Catalog No. 501060530
Encompass_Preferred
Calibration kit for validating the photometric accuracy in the range of 400-700nm and the wavelength accuracy in the range of 241-637nm for a UV/VIS spectrophotometer• Contains three neutral density filters- HF-0.2 (60%T), HF-0.5 (30%T) and HF-1.0 (10%T) for validating photometric accuracy• Contains one wavelength accuracy filter- WAV-1 UV/VIS• Contains an A30 blank and a G6 cuvette case• External dimensions of filters is 12.5x12.5x45mm• Filters are NIST traceable and produced in an ISO 17025 lab• Recommended recalibration period of 1 year
Catalog No. 50-106-0530 Supplier Fireflysci Supplier No. CALIBRATIONKIT1
Calibration kit for validating the photometric accuracy in the range of 400-700nm and the wavelength accuracy in the range of 241-637nm for a UV/VIS spectrophotometer• Contains three neutral density filters- HF-0.2 (60%T), HF-0.5 (30%T) and HF-1.0 (10%T) for validating photometric accuracy• Contains one wavelength accuracy filter- WAV-1 UV/VIS• Contains an A30 blank and a G6 cuvette case• External dimensions of filters is 12.5x12.5x45mm• Filters are NIST traceable and produced in an ISO 17025 lab• Recommended recalibration period of 1 year