Learn More
Fireflysci WAV-1 UV-VIS Advanced Holmium Oxide Wavelength Accuracy Calibration Standard (241.5-637.5nm)
SDP
Catalog No. 501060517
Encompass_Preferred
The WAV-1 UV/VIS allows calibration of wavelength accuracy in the UV and VIS range• FireflySci offers the only holmium glass reference standard in the world that has the wavelength point of 241.5nm for verifications related to DNA measurements• The following NIST wavelength peaks are provided for validating spectrophotometers: 241.5, 279.3, 287.6, 333.8, 360.8, 385.8, 418.5, 453.4, 459.9, 536.4 and 637.5nm• Uncertainty of +/- 0.2nm• External dimensions of filter is 12.5x12.5x45mm• Filter is NIST traceable and produced in an ISO 17025 lab• Recommended recalibration period of 2 years
Catalog No. 50-106-0517 Supplier Fireflysci Supplier No. WAV1 UV/VIS
The WAV-1 UV/VIS allows calibration of wavelength accuracy in the UV and VIS range• FireflySci offers the only holmium glass reference standard in the world that has the wavelength point of 241.5nm for verifications related to DNA measurements• The following NIST wavelength peaks are provided for validating spectrophotometers: 241.5, 279.3, 287.6, 333.8, 360.8, 385.8, 418.5, 453.4, 459.9, 536.4 and 637.5nm• Uncertainty of +/- 0.2nm• External dimensions of filter is 12.5x12.5x45mm• Filter is NIST traceable and produced in an ISO 17025 lab• Recommended recalibration period of 2 years