Learn More
Hach Company Stablcal Reference Suspension Set w/Cells
Supplier:  Hach Company 2897100
Encompass
Stabilised Formazine reference suspensions for the determination of lucidity/opalescence.
Catalog No. NC1497227