Learn More
Hach Company Radiometer Analytical REF201 Red Rod Reference Electrode (banana plug)
Catalog No. 501997594 Shop All Hach Company Products
Encompass
The Radiometer REF201 Reference Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl), and porous pin junction. The REF201 reference probe is recommended for general purpose measurements. The REF201 has a 1 meter fixed cable with banana plug. It can be used with Radiometer MeterLab and Hach Sension+ Laboratory Meters.
Catalog No. 50-199-7594 Supplier Hach Company Supplier No. E21M009
The Radiometer REF201 Reference Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl), and porous pin junction. The REF201 reference probe is recommended for general purpose measurements. The REF201 has a 1 meter fixed cable with banana plug. It can be used with Radiometer MeterLab and Hach Sension+ Laboratory Meters.