Learn More
Cayman Chemical RefametInIb 10mg
SDP
Catalog No. 502965928
Encompass_Preferred
An allosteric, selective inhibitor of MEK1 and MEK2 (IC50s = 19 and 47 nM, respectively); blocks phosphorylation of ERK1/2 across several human cancer cell lines differing in tissue origin and BRAF mutational status (EC50s = 2.5-16 nM); orally available and active in human tumor xenograft models
Catalog No. 50-296-5928 Supplier Cayman Chemical Supplier No. 1699710 MG
An allosteric, selective inhibitor of MEK1 and MEK2 (IC50s = 19 and 47 nM, respectively); blocks phosphorylation of ERK1/2 across several human cancer cell lines differing in tissue origin and BRAF mutational status (EC50s = 2.5-16 nM); orally available and active in human tumor xenograft models