Learn More
Medchemexpress LLC Refametinib | 923032-37-5 | MFCD18633256 | 99.8% | 572.34 g·mol⁻¹ | C19H20F3IN2O5S | 5 MG
SDP
Catalog No. 502030551
Encompass_Preferred
Refametinib is an orally active, non-ATP-competitive allosteric inhibitor of MEK1 and MEK2 used in research to modulate MAPK/ERK signaling. It exhibits low-nanomolar potency and is supplied as a high-purity solid for in vitro and preclinical studies.
  • Allosteric MEK1/MEK2 inhibitor with low-nanomolar potency.
  • Reported IC50: MEK1 = 19 nM; MEK2 = 47 nM.
  • High purity suitable for research applications.
  • Solid powder form for convenient dosing and preparation.
  • Recommended storage at low temperatures to preserve stability.
Catalog No. 50-203-0551 Supplier Medchemexpress LLC Supplier No. HY146915MG
Refametinib is an orally active, non-ATP-competitive allosteric inhibitor of MEK1 and MEK2 used in research to modulate MAPK/ERK signaling. It exhibits low-nanomolar potency and is supplied as a high-purity solid for in vitro and preclinical studies.
  • Allosteric MEK1/MEK2 inhibitor with low-nanomolar potency.
  • Reported IC50: MEK1 = 19 nM; MEK2 = 47 nM.
  • High purity suitable for research applications.
  • Solid powder form for convenient dosing and preparation.
  • Recommended storage at low temperatures to preserve stability.