Learn More
TARGETMOL CHEMICALS INC REFAMETINIB 5MG
SDP
Catalog No. 502576483
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Refametinib (RDEA119) (RDEA119 Bay 86-9766) is an effective ATP non-competitive and specific inhibitor of MEK1/2 (IC50: 19/47 nM). purity: 99%
Catalog No. 50-257-6483 Supplier TARGETMOL CHEMICALS INC Supplier No. T66365MG
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. Refametinib (RDEA119) (RDEA119 Bay 86-9766) is an effective ATP non-competitive and specific inhibitor of MEK1/2 (IC50: 19/47 nM). purity: 99%