Learn More
World Precision Instrument Reference Cells-RC1T
SDP
Catalog No. 50821932
Encompass
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
Catalog No. 50-821-932 Supplier World Precision Instrument Supplier No. RC1T
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.