Learn More
World Precision Instrument Reference Cells-RC6
SDP
Catalog No. 502521056
Encompass
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
Catalog No. 50-252-1056 Supplier World Precision Instrument Supplier No. RC6
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.