Learn More
YSI 3P LifeScience REFERENCE ELECTRODE
Catalog No. NC1152968
Encompass
This is the reference electrode cell used with the OI Analytical Cyanide by Gas Diffusion and Amperometric Flowcell. This is an easy to change, disposable electrode that produces great results with no need to fill or polish. This new version is the next generation that has a protrusion that gives better contact with the flow of base reagent producing much better low level results.
Catalog No. NC1152968 Supplier YSI 3P LifeScience Supplier No. 329513
This is the reference electrode cell used with the OI Analytical Cyanide by Gas Diffusion and Amperometric Flowcell. This is an easy to change, disposable electrode that produces great results with no need to fill or polish. This new version is the next generation that has a protrusion that gives better contact with the flow of base reagent producing much better low level results.