Learn More
EZKEM Reference Electrode, FS3700 & 9310
SDP
Catalog No. NC1813189
Encompass_Preferred
EZkem’s #329513 Reference Cell Electrode is used for the FS3700 and 9310 platforms. This is the reference electrode cell used with the OI Analytical Cyanide by Gas Diffusion and Amperometric Flowcell. This is an easy to change, disposable electrode that produces great results with no need to fill or polish. This new version is the next generation that has a protrusion that gives better contact with the flow of base reagent producing much better low level results.
Catalog No. NC1813189 Supplier EZKEM Supplier No. 329513
EZkem’s #329513 Reference Cell Electrode is used for the FS3700 and 9310 platforms. This is the reference electrode cell used with the OI Analytical Cyanide by Gas Diffusion and Amperometric Flowcell. This is an easy to change, disposable electrode that produces great results with no need to fill or polish. This new version is the next generation that has a protrusion that gives better contact with the flow of base reagent producing much better low level results.