Learn More
Hach Company Radiometer Analytical REF361 Reference Electrode (Ag/AgCl reference, sleeve junction, BNC)
Catalog No. 501997593 Shop All Hach Company Products
Encompass
The Radiometer REF361 Reference Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl), and glass sleeve junction. The REF361 reference probe is recommended for measurements in non-aqueous media or difficult samples. The REF361 has a 1 meter fixed cable with shielded BNC connector.
Catalog No. 50-199-7593 Supplier Hach Company Supplier No. E21M003
The Radiometer REF361 Reference Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl), and glass sleeve junction. The REF361 reference probe is recommended for measurements in non-aqueous media or difficult samples. The REF361 has a 1 meter fixed cable with shielded BNC connector.