Learn More
Hach Company Reference Filling Solution 1M
Supplier:  Hach Company 28564026
Encompass
Filling Solution-Reference-1 M KNO -59 mLFilling solution for electrode reference element Potassium Nitrate (KNO ) 1 M 59 mL bottle
Catalog No. 50-002-06984