Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reference Material for Flash Point Certified by The Japan Petroleum Institute, C10 Tetramer, TCI America™
SDP

Catalog No. S0556250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0556250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0556250ML Supplier TCI America Supplier No. S0556250ML
Add to Cart
Add to Cart

Chemical Identifiers

Synonym C10 Tetramer

Specifications

Color Colorless
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Reference Material for Flash Point Certified by The Japan Petroleum Institute, C10 Tetramer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.