Learn More
Thermo Scientific Soil Reference Material 1 Pack
Supplier: Thermo Scientific 33840026
Soil reference material NCS is a homogeneous, stable sample used as a calibration and quality control standard for soil analysis to enable instrument calibration, test method verification, and monitoring of accuracy and precision.