Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reference Material of Nitrogen in Residual Fuel Oil Certified by The Japan Petroleum Institute,TCI America™
SDP

Catalog No. S025120ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S025120ML 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S025120ML Supplier TCI America Supplier No. S025120ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrogen Content 0.1mass% level

Chemical Identifiers

MDL Number MFCD00197837

Specifications

Quantity 20 mL
Chemical Name or Material Reference Material of Nitrogen in Residual Fuel Oil Certified by The Japan Petroleum Institute, Nitr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.