Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reference Material of Sulfur in Residual Fuel Oil Certified by The Japan Petroleum Institute, TCI America™
SDP

Catalog No. S0225100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0225100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0225100ML Supplier TCI America Supplier No. S0225100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfur Content 0.5mass% level

Chemical Identifiers

MDL Number MFCD00197838

Specifications

Quantity 100 mL
Chemical Name or Material Reference Material of Sulfur in Residual Fuel Oil Certified by The Japan Petroleum Institute, Sulfur
EINECSNumber (9)-1689&(9)-1703&(9)-1706
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.