Learn More
Malvern Panalytical Reference Material Test Kit (M
Catalog No. NC2066118
Encompass
Reference Material Test Kit VP CAP,PEAQ DSC Manual and Auto. This kit allows users to test instrument performance. Expect material for single use, triplicate testing. This kit will ship with at least 90 days of use before material expiry.
Catalog No. NC2066118 Supplier Malvern Panalytical Supplier No. 10044946
Reference Material Test Kit VP CAP,PEAQ DSC Manual and Auto. This kit allows users to test instrument performance. Expect material for single use, triplicate testing. This kit will ship with at least 90 days of use before material expiry.