Learn More
Testo Analyzed reference oil for 270 oil tester - for calibration and adjustment of the cooking oil tester, 100 ml bottle
Catalog No. NC2074136
Encompass_Preferred
This item has a minimum order quantity of 6 bottles per supplier requirements.
Reference oil for regular checking of measuring instrument accuracy. Plant-based for calibration and adjustment of Testo cooking oil testers. Is heated to +50°C for calibration and adjustment. Analyzed reference oil for 270 oil tester, 100 ml bottle
Catalog No. NC2074136 Supplier Testo Supplier No. 0554 2650
This item has a minimum order quantity of 6 bottles per supplier requirements.
Reference oil for regular checking of measuring instrument accuracy. Plant-based for calibration and adjustment of Testo cooking oil testers. Is heated to +50°C for calibration and adjustment. Analyzed reference oil for 270 oil tester, 100 ml bottle