Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EP Reference Solution B9, Brown Reference Solution, Ricca Chemical
SDP

Catalog No. 288094 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
288094 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 288094 Supplier Ricca Chemical Company Supplier No. 2880.94
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Reference solution B9
Packaging Amber Glass Bottle
CAS 10025-77-1, 7647-01-0, 7732-18-5, 7758-99-8, 7791-13-1
Color Brown
pH Acidic
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Brown reference solution
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 120 mL
Grade EP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.