Learn More
Lovibond™ Reference Standard Kit Chlorine - 0.5 and 2.0 mg/L (MD/PM 600 series)
Catalog No. 09051502 Shop All Tintometer Group Products
Change view
Click to view available options
Wavelength Range:
530 nm
Catalog No. Wavelength Range
09-051-502 530 nm
1 options
Catalog No. 09-051-502 Supplier Lovibond™ Supplier No. 215635
Add to Cart
Add to Cart
Includes a vial to zero your instrument, 0.5mg/L and 2.0 mg/L chlorine standards for use with the PM and MD600 series of instruments.
  • Only for use with tablet methods
  • Includes reference standard vials, case, and test certificate
Specifications
Type Chlorine-DPD Free, Combined and Total
Wavelength Range 530 nm