Learn More
Lovibond™ Reference Standard Kit Chlorine
Catalog No. 09051501 Shop All Tintometer Group Products
Change view
Click to view available options
Wavelength Range:
530 nm
Catalog No. Wavelength Range
09-051-501 530 nm
1 options
Catalog No. 09-051-501 Supplier Lovibond™ Supplier No. 275650
Add to Cart
Add to Cart
Verify that your instrument is taking accurate measurements with the Reference Standard Kit Chlorine - 0.2 and 1.0 mg/L (MD100).
  • Only for instruments with tablet/ liquid reagent methods
  • Contains test certificate, zero vial, 0.2mg/L standard, 1.0 mg/L standard
Specifications
Type Chlorine-DPD Free, Combined and Total
Wavelength Range 530 nm