Learn More
USA Scientific Inc 10 ul XL TipOne filter tip refill starter pack, sterile
Catalog No. NC1030930
Encompass
TipOne 10 ul XL ultra micro filter tip refill starter pack. Includes one box with ten racks of filter tips and one box of ten filter tip refill cassettes (total 1920 tips). Sterile.
Catalog No. NC1030930 Supplier USA Scientific Inc Supplier No. 11203510
TipOne 10 ul XL ultra micro filter tip refill starter pack. Includes one box with ten racks of filter tips and one box of ten filter tip refill cassettes (total 1920 tips). Sterile.