Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte Refill for Topsoil Tour
SDPSCI_ED

Catalog No. S89509 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
4 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89509 4 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89509 Supplier LaMotte Supplier No. R5973
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent refill for use with the digital version of the LaMotte Topsoil Tour.

  • Refill of consumable items provided in original kit

Specifications

Grade 4 to 8
Age 9 to 14
For Use With (Equipment) LaMotte Topsoil Tour
Class Size 1 Class
Product Type Reagent
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.