Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Refill for Urinalysis Using Simulated Urine Kit
GREENER_CHOICESCI_ED

Catalog No. s37732 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37732 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37732 Supplier Innovating Science™ Supplier No. IS3008REF
Only null left
Add to Cart
Add to Cart

Includes four simulated replacement urine samples.

  • Includes 4 x 250 mL each of simulated urine samples: Patient X, Patient Y, Patient Z, Control

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Equipment) Urinalysis Using Simulated Urine Kit
Includes 4 x 250 mL each of simulated urine samples: Patient X, Patient Y, Patient Z, Control
Quantity 1
Dimensions (L x W x H) 5.5 x 4.25 x 6.75 in.
Class Size 15 Lab Groups
Product Type Replacement Urine Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.