Learn More
Electron Microscopy Sciences View-Pack™ Refill
SDP
Catalog No. 50931179
Encompass_Preferred
Each page holds 16 slides 3x1" (75x25mm) in individual pockets with a white back and clear front. Flap in the center keeps slides from falling out. Fold the page at the center crease to retrieve slides. 8.5" x 10.5" (22x27cm).
Catalog No. 50-931-179 Supplier Electron Microscopy Sciences Supplier No. 7154450
Each page holds 16 slides 3x1" (75x25mm) in individual pockets with a white back and clear front. Flap in the center keeps slides from falling out. Fold the page at the center crease to retrieve slides. 8.5" x 10.5" (22x27cm).