Learn More
Alkali Scientific Refillable Double Junction pH Electrode
SDP
Supplier:  Alkali Scientific LES1043B
Encompass_Preferred
The refillable double junction pH electrode is designed for accurate and stable pH measurement in laboratory and research environments. Featuring a double junction reference system, this electrode helps minimize contamination and improves measurement reliability when working with complex or challenging samples. The refillable design allows users to replenish the internal reference solution, extending the electrode’s service life and maintaining consistent performance over time. This pH electrode is suitable for use in chemical analysis, environmental testing, water quality monitoring, and general laboratory applications where dependable pH readings are essential. Its durable construction supports routine laboratory use while maintaining measurement stability and precision. The electrode is compatible with a wide range of pH meters and laboratory testing setups, making it a practical choice for laboratories that require reliable and repeatable pH measurements du
Catalog No. 50-006-61990