Learn More
Hach Company IntelliCAL PHC745 Laboratory RedRod Refillable Glass pH Electrode Kit with Calibration and Maintenance Reagents
Catalog No. NC2609515 Shop All Hach Company Products
Encompass
The IntelliCAL PHC745 kit pairs a laboratory-grade refillable glass pH electrode with a calibration and maintenance reagents pack to simplify routine calibrations and maintenance for challenging or clogging media. The electrode is designed to deliver stable, repeatable pH measurements across a wide temperature range and connects to HQD-compatible meters.
  • Refillable glass electrode suitable for clogging and dirty media.
  • Encapsulated reference element and saturated KCl filling solution for stable measurements and reduced junction clogging.
  • Includes 500 mL color-coded buffer solutions (pH 4.01, pH 7.00, pH 10.01), electrode cleaning solution, and storage solution.
  • Provides accuracy to ±0.01 pH and a measurement range of 0-14 pH.
  • 1 m cable with HQD-compatible connector
  • Operating temperature range -10 to 100 °C for broad laboratory use.
  • IP65 water resistance and 12-month warranty for reliable performance.
Catalog No. NC2609515 Supplier Hach Company Supplier No. PHC74501AP
The IntelliCAL PHC745 kit pairs a laboratory-grade refillable glass pH electrode with a calibration and maintenance reagents pack to simplify routine calibrations and maintenance for challenging or clogging media. The electrode is designed to deliver stable, repeatable pH measurements across a wide temperature range and connects to HQD-compatible meters.
  • Refillable glass electrode suitable for clogging and dirty media.
  • Encapsulated reference element and saturated KCl filling solution for stable measurements and reduced junction clogging.
  • Includes 500 mL color-coded buffer solutions (pH 4.01, pH 7.00, pH 10.01), electrode cleaning solution, and storage solution.
  • Provides accuracy to ±0.01 pH and a measurement range of 0-14 pH.
  • 1 m cable with HQD-compatible connector
  • Operating temperature range -10 to 100 °C for broad laboratory use.
  • IP65 water resistance and 12-month warranty for reliable performance.