Learn More
Grainger Refl. Letter Label P Glow in Dark PK10
Catalog No. 502753215
Encompass_Preferred
The 14R197 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing, sleeve bearing electric motor manufactured by Baldor-Reliance. It has a 1/2" diameter shaft with a 1/4" keyway and counterclockwise rotation when looking at the shaft end. The motor is totally enclosed, fan cooled with a cast iron frame and end brackets. It can operate on 50/60 Hz power and has a service factor of 1.15. The 14R197 weighs approximately 22 lbs and has standard NEMA mounting dimensions. This general purpose motor is suitable for industrial, commercial and agricultural applications.
Catalog No. 50-275-3215 Supplier Grainger Supplier No. 14R197
The 14R197 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing, sleeve bearing electric motor manufactured by Baldor-Reliance. It has a 1/2" diameter shaft with a 1/4" keyway and counterclockwise rotation when looking at the shaft end. The motor is totally enclosed, fan cooled with a cast iron frame and end brackets. It can operate on 50/60 Hz power and has a service factor of 1.15. The 14R197 weighs approximately 22 lbs and has standard NEMA mounting dimensions. This general purpose motor is suitable for industrial, commercial and agricultural applications.