Learn More
Grainger Refl. Letter Label Z Glow in Dark PK10
Catalog No. 502751937
Encompass_Preferred
The Grainger 14R208 is a 14 inch diameter, 208 volt, 1 phase, 60 Hz, 3450 RPM, 1/3 HP, 56C frame, open drip proof motor. It has a cast iron frame construction with ball bearings and a thermally protected enclosure. The motor is CSA certified and suitable for industrial, commercial, and agricultural applications. With a full load current of 5.8 Amps, this electric motor can provide reliable power transmission for pumps, fans, conveyors, compressors, and other machinery.
Catalog No. 50-275-1937 Supplier Grainger Supplier No. 14R208
The Grainger 14R208 is a 14 inch diameter, 208 volt, 1 phase, 60 Hz, 3450 RPM, 1/3 HP, 56C frame, open drip proof motor. It has a cast iron frame construction with ball bearings and a thermally protected enclosure. The motor is CSA certified and suitable for industrial, commercial, and agricultural applications. With a full load current of 5.8 Amps, this electric motor can provide reliable power transmission for pumps, fans, conveyors, compressors, and other machinery.