Learn More
HANNA INSTRUMENTS INC Refillable Double Junction pH Electrode with BNC Connector For Fluoride Applications
SDP
Catalog No. NC1655123
Encompass
The HI1143B is a glass body, refillable, double junction pH electrode with a BNC connector. This electrode has a single ceramic junction in the outer reference cell and the pH sensing portion is made with fluoride resistant glass. This design consideration is ideal for samples with fluoride.Refillable ElectrodeDouble Junction DesignFluoride Resistant Glass
Catalog No. NC1655123 Supplier HANNA INSTRUMENTS INC Supplier No. HI1143B
The HI1143B is a glass body, refillable, double junction pH electrode with a BNC connector. This electrode has a single ceramic junction in the outer reference cell and the pH sensing portion is made with fluoride resistant glass. This design consideration is ideal for samples with fluoride.Refillable ElectrodeDouble Junction DesignFluoride Resistant Glass