Learn More
MSC Reflective Apparel Safety Vest, 5-point Breakaway, ANSI 2, 1 ea
Catalog No. 5000786868
Encompass
This ANSI II Compliant Hi-Vis Safety Economy Vest features a 5-point breakaway design, providing enhanced safety in hazardous environments. Made from a lightweight and comfortable mesh fabric, it includes an internal pocket and a hook and loop front closure for ease of use. The durable 3.1 oz polyester mesh fabric helps keep the wearer cool.
  • Retro reflective tape measures 2 in. wide with 360 degree coverage
  • High visibility lime color
  • Silver fabric reflective material
  • Constructed from 100% mesh polyester
  • Breakaway design features 5 points at shoulders, sides, and front
  • Drop shoulders for comfort
  • Internal pocket for storage
Catalog No. 50-007-86868 Supplier MSC Supplier No. 38580254
This ANSI II Compliant Hi-Vis Safety Economy Vest features a 5-point breakaway design, providing enhanced safety in hazardous environments. Made from a lightweight and comfortable mesh fabric, it includes an internal pocket and a hook and loop front closure for ease of use. The durable 3.1 oz polyester mesh fabric helps keep the wearer cool.
  • Retro reflective tape measures 2 in. wide with 360 degree coverage
  • High visibility lime color
  • Silver fabric reflective material
  • Constructed from 100% mesh polyester
  • Breakaway design features 5 points at shoulders, sides, and front
  • Drop shoulders for comfort
  • Internal pocket for storage