Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Reflective Floor Tape

Catalog No. 19805178 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-805-178 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-178 Supplier Accuform Signs Supplier No. PTM802WT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made of engineer grade material with bounceback reflectivity

  • Improved daytime, low-light, and nighttime visibility
  • Permanent adhesive with peel-off liner
  • Suitable for outdoor use on poles, barricades, fences, buildings, private roads, and off-road vehicles
  • 5.5-mil thick urethane tape

Specifications

Color White
For Use With (Application) Suitable for outdoor use on poles, barricades, fences, buildings, private roads, and off-road vehicles
Length (English) 30 ft.
Thickness 5.5 mil
Width (English) 2 in.
Adhesive Type Permanent Adhesive
Material Urethane
Type Reflective Floor Tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.