Learn More
Grainger Reflective Numbers And Letters G PK25
Catalog No. 502750398
Encompass_Preferred
The PK-25-1VE19 is a 1/4 HP, 115V, single phase, open drip proof, sleeve bearing, electric motor made by Century Electric Motors. It has a totally enclosed fan cooled design with a 1.15 service factor. The motor operates at 1725 RPM with a full load amp rating of 3.6 Amps. It features a 48 frame size with a standard C-face mounting. The motor dimensions are 6-1/2" diameter x 7-3/4" long and it weighs 15 lbs.
Catalog No. 50-275-0398 Supplier Grainger Supplier No. 14V783
The PK-25-1VE19 is a 1/4 HP, 115V, single phase, open drip proof, sleeve bearing, electric motor made by Century Electric Motors. It has a totally enclosed fan cooled design with a 1.15 service factor. The motor operates at 1725 RPM with a full load amp rating of 3.6 Amps. It features a 48 frame size with a standard C-face mounting. The motor dimensions are 6-1/2" diameter x 7-3/4" long and it weighs 15 lbs.