Learn More
Grainger Reflective Numbers And Letters X PK25
Catalog No. 502752224
Encompass_Preferred
The Brady Number Label is a 1 inch character height label made of vinyl with a permanent acrylic adhesive. It is designed for indoor use on smooth, flat surfaces like metal, plastic, glass, and painted wood. The label has high contrast black numbers on a white background for easy visibility. It can withstand temperatures from -40°F to 200°F. The label comes in sheets with 30 labels per sheet. It meets UL969 and CSA C22.2 No. 0.15 standards.
Catalog No. 50-275-2224 Supplier Grainger Supplier No. 14V801
The Brady Number Label is a 1 inch character height label made of vinyl with a permanent acrylic adhesive. It is designed for indoor use on smooth, flat surfaces like metal, plastic, glass, and painted wood. The label has high contrast black numbers on a white background for easy visibility. It can withstand temperatures from -40°F to 200°F. The label comes in sheets with 30 labels per sheet. It meets UL969 and CSA C22.2 No. 0.15 standards.