Learn More
Grainger Reflective Numbers And Letters 6 PK25
Catalog No. 502752953
Encompass_Preferred
The Stranco Inc. Reflective Number Label 6 is a reflective label that can be used for identification and inventory control purposes. It measures 6 inches tall and is made of a reflective vinyl material with pressure sensitive adhesive backing. The label has bold black numbers that reflect light back to the source for visibility in low light conditions. It can adhere to clean, flat surfaces and withstand outdoor and industrial environments. This reflective label provides an easy way to identify items, parts or locations that need high visibility numbering.
Catalog No. 50-275-2953 Supplier Grainger Supplier No. 14V810
The Stranco Inc. Reflective Number Label 6 is a reflective label that can be used for identification and inventory control purposes. It measures 6 inches tall and is made of a reflective vinyl material with pressure sensitive adhesive backing. The label has bold black numbers that reflect light back to the source for visibility in low light conditions. It can adhere to clean, flat surfaces and withstand outdoor and industrial environments. This reflective label provides an easy way to identify items, parts or locations that need high visibility numbering.