Learn More
Grainger Reflective Numbers And Letters T PK25
Catalog No. 502753360
Encompass_Preferred
The Brady Letter Label is a 2.5 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They feature legible lettering that is easy to read from a distance. The vinyl material is durable in indoor and outdoor environments. Overall, these letter labels provide a versatile and long-lasting solution for clear identification and labeling in various applications.
Catalog No. 50-275-3360 Supplier Grainger Supplier No. 14R238
The Brady Letter Label is a 2.5 inch character height label with a white background and black text. It is made of vinyl material and has an adhesive backing for application. The labels are useful for identifying wires, panels, components, equipment, and more. They feature legible lettering that is easy to read from a distance. The vinyl material is durable in indoor and outdoor environments. Overall, these letter labels provide a versatile and long-lasting solution for clear identification and labeling in various applications.