Learn More
Grainger Reflective Numbers And Letters 0 PK25
Catalog No. 502751509
Encompass_Preferred
The Brady Number Label is a 2.5" x 1" label with 2.5" tall characters for easy visibility. It is made of vinyl cloth material and features permanent acrylic adhesive for application on a variety of surfaces. The label can display up to 6 digits or letters in black print on a white background. It is suitable for indoor or outdoor use and rated for temperatures from -40°F to 160°F. The label comes in a pack of 250.
Catalog No. 50-275-1509 Supplier Grainger Supplier No. 14R209
The Brady Number Label is a 2.5" x 1" label with 2.5" tall characters for easy visibility. It is made of vinyl cloth material and features permanent acrylic adhesive for application on a variety of surfaces. The label can display up to 6 digits or letters in black print on a white background. It is suitable for indoor or outdoor use and rated for temperatures from -40°F to 160°F. The label comes in a pack of 250.