Learn More
Grainger Reflective Numbers And Letters P PK25
Catalog No. 502751610
Encompass_Preferred
The Stranco Inc. Reflective Letter Label 2-8CL50 is a 2 inch tall reflective letter label made of durable acrylic plastic with pressure sensitive adhesive backing. It has a service temperature range of -40°F to 200°F. The label has a high gloss finish and comes in black color with silver reflective lettering reading "50". It meets DOT C2 reflective requirements and is ideal for identifying equipment for night visibility. The label can adhere to clean flat surfaces like metal, glass, plastic, painted and powder coated surfaces. It resists UV rays, abrasion, chemicals and moisture. The label comes in a pack of 10.
Catalog No. 50-275-1610 Supplier Grainger Supplier No. 14V792
Add to Cart
Add to Cart
The Stranco Inc. Reflective Letter Label 2-8CL50 is a 2 inch tall reflective letter label made of durable acrylic plastic with pressure sensitive adhesive backing. It has a service temperature range of -40°F to 200°F. The label has a high gloss finish and comes in black color with silver reflective lettering reading "50". It meets DOT C2 reflective requirements and is ideal for identifying equipment for night visibility. The label can adhere to clean flat surfaces like metal, glass, plastic, painted and powder coated surfaces. It resists UV rays, abrasion, chemicals and moisture. The label comes in a pack of 10.