Learn More
Grainger Reflective Numbers And Letters 5 PK25
Catalog No. 502748697
Encompass_Preferred
The Brady Number Label is a 2.5" x 1" label with 14 characters per line. It is made of white polyester with a permanent acrylic adhesive and has a temperature range of -40°F to 302°F. The label can be used for wire and cable marking, panel and equipment ID, barcode labeling, and more. It has high adhesion and is resistant to chemicals, abrasion, and UV light. The label comes in a pack of 250.
Catalog No. 50-274-8697 Supplier Grainger Supplier No. 14R214
The Brady Number Label is a 2.5" x 1" label with 14 characters per line. It is made of white polyester with a permanent acrylic adhesive and has a temperature range of -40°F to 302°F. The label can be used for wire and cable marking, panel and equipment ID, barcode labeling, and more. It has high adhesion and is resistant to chemicals, abrasion, and UV light. The label comes in a pack of 250.