Learn More
Grainger Reflective Numbers And Letters M PK25
Catalog No. 502749620
Encompass_Preferred
The Brady Letter Label is a 2.5 inch tall letter label with a white background and black text. It has a permanent acrylic adhesive and is made from vinyl. This label is suitable for indoor or outdoor use on a variety of surfaces. It can withstand temperatures from -40°F to 200°F. The label comes in a pack of 250 and has a height of 2.5 inches and width of 1 inch. It meets the BS5609 certification for durability and performance. This is an ideal letter label for identifying wires, panels, equipment and more.
Catalog No. 50-274-9620 Supplier Grainger Supplier No. 14R231
The Brady Letter Label is a 2.5 inch tall letter label with a white background and black text. It has a permanent acrylic adhesive and is made from vinyl. This label is suitable for indoor or outdoor use on a variety of surfaces. It can withstand temperatures from -40°F to 200°F. The label comes in a pack of 250 and has a height of 2.5 inches and width of 1 inch. It meets the BS5609 certification for durability and performance. This is an ideal letter label for identifying wires, panels, equipment and more.