Learn More
Grainger Reflective Numbers And Letters S PK25
Catalog No. 502749813
Encompass_Preferred
The Brady Number Label is a 2.5" x 1" label with 14 characters per line. It is made of white vinyl material with black print. The label has a permanent acrylic adhesive and is designed for indoor use on smooth, flat surfaces like plastic, metal, and painted wood. It can withstand temperatures from -40°F to 200°F. The label comes in a pack of 250 and has a catalog number of 14R237. It can be used for basic product identification and inventory management.
Catalog No. 50-274-9813 Supplier Grainger Supplier No. 14R237
The Brady Number Label is a 2.5" x 1" label with 14 characters per line. It is made of white vinyl material with black print. The label has a permanent acrylic adhesive and is designed for indoor use on smooth, flat surfaces like plastic, metal, and painted wood. It can withstand temperatures from -40°F to 200°F. The label comes in a pack of 250 and has a catalog number of 14R237. It can be used for basic product identification and inventory management.