Learn More
Grainger Reflective Numbers And Letters 4 PK25
Catalog No. 502752049
Encompass_Preferred
The Brady Number Label is a 1 inch character height label made of vinyl material. It has a white background with black print and is suitable for indoor or outdoor use. The label has an adhesive back and comes in sheets of 30 labels. It can be used for identifying wires, panels, components, equipment and more. The temperature range is -40°F to 176°F.
Catalog No. 50-275-2049 Supplier Grainger Supplier No. 14V808
The Brady Number Label is a 1 inch character height label made of vinyl material. It has a white background with black print and is suitable for indoor or outdoor use. The label has an adhesive back and comes in sheets of 30 labels. It can be used for identifying wires, panels, components, equipment and more. The temperature range is -40°F to 176°F.