Learn More
Grainger Reflective Numbers And Letters U PK25
Catalog No. 502748693
Encompass_Preferred
The PK-25-1VE19 is a 1/4 HP, 115V, single phase, open drip proof, sleeve bearing electric motor manufactured by Century Electric Motors. It operates at 1725 RPM with a 1/4" diameter shaft and includes a 6 foot power cord. The motor is thermally protected, has manual reset overload protection, and is CSA certified. With a 56C frame, it can provide 0.19 horsepower and is commonly used for driving fans, blowers, and pumps in light industrial applications. The motor dimensions are 3-1/2" diameter x 5-7/16" long and it weighs approximately 6 pounds.
Catalog No. 50-274-8693 Supplier Grainger Supplier No. 14V797
The PK-25-1VE19 is a 1/4 HP, 115V, single phase, open drip proof, sleeve bearing electric motor manufactured by Century Electric Motors. It operates at 1725 RPM with a 1/4" diameter shaft and includes a 6 foot power cord. The motor is thermally protected, has manual reset overload protection, and is CSA certified. With a 56C frame, it can provide 0.19 horsepower and is commonly used for driving fans, blowers, and pumps in light industrial applications. The motor dimensions are 3-1/2" diameter x 5-7/16" long and it weighs approximately 6 pounds.