Learn More
Grainger Reflective Numbers And Letters Q PK25
Catalog No. 502753059
Encompass_Preferred
The Brady Letter Label is a 2.5 inch tall label with 14 characters per inch. It is made of white vinyl material and has a permanent acrylic adhesive backing. The labels are designed for indoor use on flat, smooth surfaces like file folders, shelves, and drawers. They can identify contents clearly and legibly. The roll contains 750 labels and measures 2.25 inches wide by 36 yards long. Brady labels resist moisture, abrasion, chemicals and sunlight. They offer a clean removal when necessary. This provides durable, long-lasting identification.
Catalog No. 50-275-3059 Supplier Grainger Supplier No. 14V793
The Brady Letter Label is a 2.5 inch tall label with 14 characters per inch. It is made of white vinyl material and has a permanent acrylic adhesive backing. The labels are designed for indoor use on flat, smooth surfaces like file folders, shelves, and drawers. They can identify contents clearly and legibly. The roll contains 750 labels and measures 2.25 inches wide by 36 yards long. Brady labels resist moisture, abrasion, chemicals and sunlight. They offer a clean removal when necessary. This provides durable, long-lasting identification.