Learn More
Grainger Reflective Numbers And Letters C PK25
Catalog No. 502751166
Encompass_Preferred
The Stranco Inc. Reflective Letter Label 2-8CL50 is a 2 inch tall reflective letter label made of durable acrylic plastic with pressure sensitive adhesive backing. It has a service temperature range of -40°F to 200°F. The label has a high gloss finish and comes in black color with silver reflective lettering reading "50". It is designed for use on electrical panels, machinery, bins, and more to identify circuits or components. This reflective label improves visibility in low light conditions. It meets ANSI Z535.1 standards. The label comes in a pack of 10.
Catalog No. 50-275-1166 Supplier Grainger Supplier No. 14V817
The Stranco Inc. Reflective Letter Label 2-8CL50 is a 2 inch tall reflective letter label made of durable acrylic plastic with pressure sensitive adhesive backing. It has a service temperature range of -40°F to 200°F. The label has a high gloss finish and comes in black color with silver reflective lettering reading "50". It is designed for use on electrical panels, machinery, bins, and more to identify circuits or components. This reflective label improves visibility in low light conditions. It meets ANSI Z535.1 standards. The label comes in a pack of 10.